Learn More
Trajan™ Thermo Scientific Instrument Ferrules
Codice prodotto. 15248883
Click to view available options
Diameter (Metric) Inside:
0.10 to 0.25 mm
Dimensione della confezione:
pezzi 10
Codice prodotto. 15248883
Marca: Trajan™ 0726549
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
SilTite FingerTite delivers an easy, leak-free installation for capillary columns without the use of any tools
  • Next generation ferrule system for gas chromatography
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifica
Tipo Detector/Injectors
Materiale Graphite, Vespel
Diameter (Metric) Inside 0.10 to 0.25 mm