Learn More
Honeywell™ SHARPFLEX NIT Gloves
Click to view available options
Size:
7
8
9
10
11
Dimensione della confezione:
1 paio
Descrizione
- Knitted glove in composite and para-aramid yarn
- The knitting technique provides the glove with excellent fit and high dexterity
- The composite yarn is reliable and safe protection
- The para-aramid yarn assure a resistance to the contact heat and cut risks
- Plated polyamide inside offers a greater comfort and limit the contact of the skin with composite fiber
- Black nitrile foam coating on palm
- Nitrile coating provides good protection against oil and grease
- The foam coating makes the glove more flexible than standard nitrile
Specifica
Specifica
| Certificazione/Conformità | EN 420-03 EEC Basic standards, EN 388-16 Mechanical hazards |
| Descrizione | Para-aramid and composite, 13 gauge |
| Colore | Green, Yellow, Black |
| Materiale | Nitrile, Polyamide |
| Size | 10 |
| Lunghezza (sistema metrico) | 25 cm |