missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sheep anti-Mouse IgG (H+L) Secondary Antibody, HRP, Novus Biologicals™
Sheep Polyclonal Antibody has been used in 2 publications
Marca: Novus Biologicals NB120-6808
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifica
| IgG | |
| Polyclonal | |
| HRP | |
| immunoglobulin heavy constant gamma 1 (G1m marker) | |
| Sheep anti-Mouse IgG (H+L) Secondary Antibody [HRP] was produced by repeated immunization with Mouse IgG whole molecule in sheep. | |
| 2 mg | |
| Secondary | |
| Reconstitute with 1.0 ml deionized water (or equivalent). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| LYOPH | |
| Western Blot 1:1000 - 1:5000, ELISA 1:100000 - 1:200000, Immunohistochemistry 1:500 - 1:2500 | |
| Sheep | |
| Multi-step | |
| RUO | |
| Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Peroxidase, anti-Sheep Serum, Mouse IgG and Mouse Serum. | |
| Mouse | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto