Learn More
Sheep anti-Rabbit IgG (H+L) Secondary Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Marca:  Novus Biologicals NBP1-75349
Codice prodotto. 18512344
  • 237.00€ / 2mg
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Rabbit samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoassay, Immunomicroscopy, Chemiluminescence Immunoassay
TRUSTED_SUSTAINABILITY
Specifica
Specifica
IgG (H+L)
Polyclonal
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoassay, Immunomicroscopy 1:10 - 1:500, Chemiluminescence Immunoassay (CIA) (CIA)
Sheep
Affinity purified
RUO
Secondary
Store at 4°C. Do not freeze.
IgG
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Unconjugated
10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2
This Sheep anti-Rabbit IgG (H+L) Secondary Antibody was developed against purified rabbit IgG (H&L).
2 mg
Adaptive Immunity, Immunology
Rabbit
Purified
Suggerimenti di prodotto
Suggerimenti di prodotto
Videos
SDS
Documenti
Documenti
Certificati
Promozioni
Promozioni