missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sheep anti-Rabbit IgG whole molecule, Texas Red, Polyclonal, Secondary Antibody, Abnova™
Marca: Abnova PAB10843.2mg
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
| Sheep anti-Rabbit IgG whole molecule | |
| Polyclonal | |
| Sheep anti-rabbit IgG recognizes rabbit IgG whole molecule. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Texas Red. | |
| Lyophilized from 10mM potassium phosphate, 75mM NaCl, 50% (v/v) glycerol, pH 7.2 (0.01% (w/v) sodium azide, 10mg/mL BSA (ImmunoglobulLyophilized from and Protease free)) | |
| Rabbit IgG whole molecule | |
| 2 mg | |
| Rabbit | |
| Lyophilized |
| Flow Cytometry, Immunofluorescence | |
| Texas Red | |
| The optimal working dilution should be determined by the end user. | |
| Sheep | |
| This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. | |
| Secondary | |
| Store at 4°C on dry atmosphere. After reconstitution with 1 mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto