missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIAH2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12651-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
SIAH-2 (seven in absentia homolog 2) is an E3 ligase that catalyzes ubiquitination and proteasome-mediated degradation of protein substrates. SIAH-2 encodes a 324 amino acid protein that shares 77% identity with human SIAH-1 and 68% identity with the Drosophila sina (seven in absentia) gene, on which development of the Drosophila R7 photoreceptor is dependent. SIAH-2 targets TRAF2 (which regulates cell responses to stress and cytokines through the regulation of key stress-signaling cascades) for degradation under stress conditions such as hypoxia. It targets HIF-1 alpha prolyl hydroxylase 3 (PHD3) for degradation upon exposure to hypoxic conditions, which coincides with an increase in SIAH-2 transcription. SIAH-2 can decrease TNF-alpha-dependent induction of JNK activity and transcriptional activation of NFkB. SIAH-2 null mice subjected to hypoxia display an impaired respiratory response and reduced levels of hemoglobin.Specifica
| SIAH2 | |
| Polyclonal | |
| Unconjugated | |
| SIAH2 | |
| hSiah2, Seven in absentia homolog 2, Siah 2, SIAH2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 140593, 20439, 6478 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O43255, Q06986, Q8R4T2 | |
| SIAH2 | |
| SIAH2 Fusion Protein Ag3342 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto