Learn More
Endecotts™ Sieve Calibration Ball
Click to view available options
Dimensione particelle:
120 mesh
14 mesh
400 mesh
missing translation for 'unitSize'
pezzi 3
pezzi 5
Description
- Spheres are traceable to the United Kingdom National Physical Laboratory
- Calibrate sieves to an accuracy of 1μm
- Soda-lime glass microspheres pass over the entire surface of the sieve and through every aperture
- Standards include a calibration chart
Specifications
Specifications
| Product Type | Microspheres |
| Materiale | Soda-lime glass |
| Da utilizzare con (apparecchiatura) | Sieve |
| Dimensione particelle | 14 mesh |