Learn More
Epredia™ Signature Series™ Clarifier™ 1, 2
Sfoglia Tutto Epredia Prodotti
Click to view available options
Quantità:
1 gal.
4 x 1 gal.
Tipo:
Clarifier 1
Clarifier 2
Dimensione della confezione:
Pezzo
pezzi 4
Descrizione
- Improves contrast between Nuclei and Cytoplasm
- Increases cell transparency
- Removes background staining caused by excessive tissue adhesive
- Removes unwanted hematoxylin staining from adhesive without affecting nuclear staining
- Clarifier 1 is recommended with Hematoxylin 7211 and Hematoxylin 1
- Clarifier 2 is recommended with Hematoxylin 7212 and Hematoxylin 2
Specifica
Specifica
| Tipo | Clarifier 2 |
| Quantità | 1 gal. |