Learn More
Epredia™ Signature Series™ Cyto-Stain™
Sfoglia Tutto Epredia Prodotti
Click to view available options
Quantità:
4 pt.
1 gal.
Dimensione della confezione:
Pezzo
pezzi 4
Descrizione
- Use for all gynecological and nongynecological procedures
- Reduces staining time
- Cost-effective, as user performs only one counterstain
- Cyto-Stain provides a full range of familiar cytoplasmic colors and control over the staining intensities
- Cyto-Stain G is a modification to produce greener cyanophilic hues in intermediate and basal cells
Specifica
Specifica
| Tipo | Cytoplasmic stain |
| Quantità | 1 gal. |