Learn More
Silica DCC, MP
Codice prodotto. 17887044 Sfoglia Tutto MP Biomedicals Prodotti
Click to view available options
:
MP Silica DCC, 25 kg
Codice prodotto. 17887044
Marca: MP Biomedicals 0204530
Dry Column Chromatography (DCC) bridges the gap between analytical Thin-Layer Chromatography (TLC) and the production-oriented column chromatography (CC)
Adsorbents for DCC imitate the chemical and physical properties of Thin-Layer Chromatography as closely as possible and can be used without any further preparation. Columns may be filled directly from the bottle in which the adsorbent was supplied.
TRUSTED_SUSTAINABILITY
Specifica
Punti di ebollizione 2230°C
Materiale o nome chimico Silicon dioxide
Punto di fusione 1710°C
CAS 7631-86-9
pH 7
Quantità 25 kg
Formula molecolare O2Si
Numero MDL MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILES O=[Si]=O
IUPAC Name silanedione
Molecular Weight (g/mol) 60.08
Densità 0.5 g/mL
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.