Learn More
DWK Life Sciences Silicone Sealing Ring, VMQ
Sfoglia Tutto DWK Life Sciences Prodotti
Click to view available options
Diameter (Metric) Inside:
11.0 to 13.0 mm
13.0 to 15.0 mm
15.0 to 17.0 mm
17.0 to 19.0 mm
25.0 to 27.0 mm
31.0 to 33.0 mm
5.5 to 6.5 mm
7.5 to 9.0 mm
9.0 to 11.0 mm
Joint Size:
GL 14
GL 18
GL 25
GL 32
GL 45
Dimensione della confezione:
pezzi 10
Descrizione
The silicone sealing ring matches the screw connection piece and has good sealing properties. Due to the PTFE layer on one side of the sealing ring, there is a certain inertness of the material. The included hole allows the insertion of glass rods, olives, hoses, etc.
- With bonded PTFE face
Specifica
Specifica
| Diameter (Metric) Inside | 25.0 to 27.0 mm |
| Da utilizzare con (applicazione) | Sealing; Connecting glass tubes; Suitable for PBT screw cap with aperture |
| Joint Size | GL 45 |
| Quantità | 10/Pk. |
| Dimensioni (sistema metrico) | Fits Tubing ID: 25.0 to 27.0 mm |
| Diametro (sistema metrico) parte esterna | 42 mm |
| Materiale | Silicone (VMQ) |