Learn More
Fisherbrand™ General Purpose Silicone Sheet
Sfoglia Tutto Fisherbrand Prodotti
Click to view available options
Dimensioni (L x P x A):
360 x 360 x 1 mm
360 x 360 x 1.5 mm
360 x 360 x 2 mm
360 x 360 x 3 mm
Dimensione della confezione:
pezzi 5
Descrizione
- Available plain or adhesive backed
- Suitable for use at -40° to +200°C and +230°C short-term
- Can be supplied in sheet form or converted into strips, seals, washers, gaskets and fabricated articles
Specifica
Specifica
| Descrizione | 3 x 360 x 360mm |
| Dimensioni (L x P x A) | 360 x 360 x 3 mm |