Learn More
KNF Neuberger SIMDOS™ 02 Diaphragm Liquid Dosing Vacuum Pump
Click to view available options
Modello:
FEM 1.02 KT.18RCP
FEM 1.10 FT.18S
Materiale:
PTFE Head
Polypropylene Head
Dimensione della confezione:
Pezzo
Descrizione
- Simple, intuitive setup and operation
- Easy to calibrate in minutes
- Runs dry without damage
- Self-priming up to 9 ft. of water
- Splash-proof IP65 housing
- Analog and impulse logic control (RC) and with RS232 (RCP)
- Small footprint conserves benchtop space
- Adaptable to viscous fluid characteristics
Specifica
Specifica
| Controllo | RS232 Remote Control |
| Dimensioni (L x P x A) | 150 x 93 x 144 mm |
| Materiale | Polypropylene Head |
| Modello | FEM 1.02 KT.18RCP |
| Portata | 0.03 to 20 mL/min. |
| Frequenza | 50/60 Hz |
| Tipo di pompa | Pompa da vuoto a diaframma |
| Valori nominali armadio | IP65 |
| Applicazioni | Pharmacy |
| Maximum Pressure | 85 psig |
| Vedi altri risultati |