Learn More
Simport™ Scientific M480 Disposable Embedding Cassettes
For holding and identifying tissue samples in processing, embedding and sectioning procedures during in vitro diagnostics
Descrizione
- Made of acetal polymer and resistant to chemical action of histological solvents
- Fit securely in microtome chuck adapters
- Accepts standard metal lids that keep specimens safe during processing
- Slanted writing surface for easy sample identification
- Available in three colors
Certifications
CE marked
Specifica
| Acetal polymer |