Learn More
Simport™ Scientific CytoSep™ Funnels for Shandon™ Cytospin 4™ Centrifuges
For use with Thermo Scientific™ Shandon Cytospin 4 Centrifuges
Specifica
| Da utilizzare con (apparecchiatura) | Shandon Cytospin™ 4 Cytocentrifuge |
|---|
Descrizione
- All plastic disposable sample chambers do not need metal clips
- Available with or without pre-attached filter card
Specifica
| Shandon Cytospin™ 4 Cytocentrifuge |