Learn More
Simport™ Scientific E-Z LOAD™ Histosette™ II Tissue Cassette Bases
For use with Micro Writer™, Shur/Mark™ and Lamb™ cassette labeling instruments
Specifica
| Tipo di chiusura | Integral Lid |
|---|---|
| Materiale | Acetal Polymer |
| Stile pori | Slots |
| Angolo della faccia di scrittura | 45° |
Descrizione
- Disposable, plastic cassettes hold tissue specimens during embedding, processing, and storage
- Molded from a special high-density acetal polymer
- Resistant to the chemical action of histological solvents
- Efficient flow-through slots maximize fluid exchange and ensure proper drainage
- The anterior writing surface is slanted at a 45° angle
- Available in 11 non-cytotoxic, non-metallic colors
Order Info
- Stacks of 50 cassettes each; case contains two boxes of 20 stacks
- Bases and covers must be ordered separately
Specifica
| Integral Lid | |
| Acetal Polymer | |
| Slots | |
| 45° |