Learn More
Simport™ Scientific SLIMSETTE™ M509SL Tissue Processing/Embedding Cassettes in QuickLoad™ Sleeves
A new generation of cassettes, prepackaged for use in automatic cassette labeling instruments
Specifica
| Tipo di chiusura | Integral Lid |
|---|---|
| Materiale | Acetal Polymer |
| Stile pori | Slots |
| Angolo della faccia di scrittura | 45° |
Descrizione
- Made of acetal polymer
- Transparent sleeve allows clear view of cassettes during printing
- Efficient flow-through slots maximize fluid exchange and ensure proper drainage
- Lid can be opened and closed as often as necessary, and will always relock securely
- Large labeling area for easy identification
- Anterior writing area at a 45° angle
Specifica
| Integral Lid | |
| Acetal Polymer | |
| Slots | |
| 45° |