Learn More
Simport™ Scientific Tamper Evident Screw Caps
For all applications requiring tamper evident seal for utmost security and sample integrity
Includes: Lip seal
Descrizione
Descrizione
  • Made of polyethylene
  • Features long skirt and super fast thread design
  • Can be removed or sealed with single turn
  • With leakproof seal