Learn More
Simport™ Scientific Tamper Evident Screw Caps
For all applications requiring tamper evident seal for utmost security and sample integrity
Includes: Lip seal
Descrizione
- Made of polyethylene
- Features long skirt and super fast thread design
- Can be removed or sealed with single turn
- With leakproof seal