Learn More
Simport™ Scientific SLIMSETTE™ M509 Tissue Processing/Embedding Cassettes
SLIMSETTE™ M509 Tissue Processing/Embedding Cassettes are a new generation of cassettes that are more compact and easier to use.
Specifica
| Quantità | 3 x 500/Cs. |
|---|---|
| Angolo della faccia di scrittura | 45° |
| Tipo di chiusura | Integral Lid |
| Materiale | Acetal Polymer |
| Product Type | Tissue Cassette |
Descrizione
- Made of acetal polymer
- Design has a unique recessed cover
- Cassettes are thinner, allowing more to be stacked in automatic labeling machines
- One-piece integral lid
- Large labeling area for easy identification
- Anterior writing area at a 45° angle
Specifica
| 3 x 500/Cs. | |
| Integral Lid | |
| Tissue Cassette |
| 45° | |
| Acetal Polymer |