Learn More
Thermo Scientific™ Single-cell sample holders for GENESYS™ and BioMate™ 160 Spectrophotometers
Codice prodotto. 15520592 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Da utilizzare con (apparecchiatura):
4-position carousel base
GENESYS 140/150/180,BioMate160
GENESYS 30/40/50
Spectronic200,GENESYS 30/40/50
Tipo:
Beckman-style Cell Holder
Cylindrical Cell Holder
Disposable Microcell Holder
Hose Entry/Exit Kit
Indiv Adjustable Filter Holder
Long Path Cell Holder
Sample Holder for Film/Filter
Single Cell Sample Holder
Test Tube Cap
Test Tube and Vial Holder
Capacità:
10 mm cuvette (1)
10 to 100 mm cuvette (1)
12 mm to 25 mm tubes and vials
20 to 100 mm cuvette (1)
5 - 100 mm cylindrical cells
5 to 100 mm cuvette (1)
5 to 100mm cylindrical cuvette
50 μL microcells, Uvettes
Beckman-style short microcells
Entry and exit hoses (incl)
Filter/lenses, 10 mm thick
Filter/lenses, 6 mm thick
Filters up to 10 mm thick
Single-cell cuvette, 10mm tube
Square 10 mm PL Cuvette
Tall tubes, vials (lid open)
Dimensione della confezione:
Pezzo
Codice prodotto. 15520592
Marca: Thermo Scientific™ 840277500
Simplify sampling with easy access
Make the most of every sample. Accessories for Thermo Scientific™ GENESYS™ UV-Vis spectrophotometers are designed to enable you to work smarter, not harder. We offer a wide variety of options to meet your measurment requirments. Cable-free, snap-in design allows for easy exchange of accessories.
We provide single-cell solutions for measuring different types of samples.
Get the most from your measurments with accessories for the GENESYS Vis/UV-Vis Spectrophotometer family. Simplify sampling with easy access, flexibility and choices to fit your varied testing needs.
TRUSTED_SUSTAINABILITY
Specifications
Descrizione Test tube and vial holder accommodating 12 mm diameter up to 25 mm diameter test tubes and vials
Tipo Test Tube and Vial Holder
Da utilizzare con (applicazione) Colorimetric/spectroscopic measurements in test tube cuvettes.
OD600 measurements for cell growth monitoring in tubes.
Monitoring cell growth in nephelo-culture flasks.
Measurements in 16 mm screw-cap vials
Da utilizzare con (apparecchiatura) GENESYS 30/40/50
Capacità 12 mm to 25 mm tubes and vials
Include One cuvette holder built into a sample compartment tray. Snaps into place magnetically. No tools required.