Learn More
Masterflex™ Single-Use Flow Tube Sensors
Click to view available options
Portata flusso d'acqua:
0.03 to 2 L/min.
0.06 to 2 L/min.
0.50 to 20 L/min.
1.50 to 40 L/min.
3 to 200 L/min.
Tipo:
Flow Sensor (Clamp-mount)
Flow Sensor (Click-mount)
Flow Sensor (Clip-mount)
Flow Sensor (Tubeholder)
Materiale:
PFA
PVDF
Dimensione della confezione:
Pezzo
Descrizione
- Modular interchangeable flow sensor systems with single-use design
- Accurate measurement readings at low flow: ±1%
- Low operating cost, disposable flow tubes for optimal contamination-free production
- Ideal for single-use applications, but also suitable for long-term measurement
- Flow tubes and turbine sensors are produced and tested under cleanroom conditions
Specifica
Specifica
| Tipo di display | None |
| Tensione | 5/24 VDC |
| Tipo | Flow Sensor (Clamp-mount) |
| Portata flusso d'acqua | 0.03 to 2 L/min. |
| Temperatura operativa max (sistema imperiale) | 176°F |
| Temperatura operativa max (sistema metrico) | 80°C |
| Materiale | PVDF |