Learn More
SIRT2 Mouse anti-Human, CoraLite 488, Clone: 1D8G10, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech CL488-66410-100UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The SIRT2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SIRT2. This antibody recognizes human, mouse, rat, pig antigen. The SIRT2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifica
| SIRT2 | |
| Monoclonal | |
| CoraLite 488 | |
| 100 μL | |
| 22933 | |
| IgG1 |
| Immunofluorescence | |
| 1D8G10 | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |