missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC1A5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-32239
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
SLC1A5 Polyclonal specifically detects SLC1A5 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifica
| SLC1A5 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| AAAT, ASCT2M7VS1, ATB(0), Baboon M7 virus receptor, M7V1ATBO, neutral amino acid transporter B, neutral amino acid transporter B(0), R16, RD114 virus receptor, RD114/simian type D retrovirus receptor, RDR, RDRCFLJ31068, Sodium-dependent neutral amino acid transporter type 2, solute carrier family 1 (neutral amino acid transporter), member 5, Solute carrier family 1 member 5 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 6510 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg lysate | |
| NP_005619.1 | |
| SLC1A5 | |
| The immunogen this antibody was made to, maps to a region between residue 450 to 500 of human Sodium-Dependent Neutral Amino Acid Transporter Type 2 using the numbering given in entry NP_005619.1 (GeneID 6510). | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto