missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC22A3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA542486
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Peptide sequence: PDHYWCRGPS AAALAERCGW SPEEEWNRTA PASRGPEPPE RRGRCQRYLL Sequence homology: Cow: 79%; Human: 100%; Pig: 86%.
Polyspecific organic cation transporters in the liver, kidney, intestine, and other organs are critical for elimination of many endogenous small organic cations as well as a wide array of drugs and environmental toxins. This gene is one of three similar cation transporter genes located in a cluster on chromosome 6. The encoded protein contains twelve putative transmembrane domains and is a plasma integral membrane protein. [provided by RefSeq, Jul 2008].
Specifica
| SLC22A3 | |
| Polyclonal | |
| Unconjugated | |
| SLC22A3 | |
| 3-Oct; EMT; EMT organic cation transporter 3; EMTH; extraneuronal monoamine extraneuronal monoamine; extraneuronal monoamine transporter; Oct3; Orct3; Organic cation transporter 3; Slc22a3; Slca22a3; solute carrier family 22 (extraneuronal monoamine transporter), member 3; solute carrier family 22 (organic cation transporter), member 3; solute carrier family 22 member 3; solute carrier family 22, member 3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 6581 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O75751 | |
| SLC22A3 | |
| Synthetic peptide directed towards the N-terminal of human SLC22A3 (aa 46-95). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto