missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC27A1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Marca: Invitrogen™ PA585002
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Immunogen sequence: QREGFDPRQT SDRLFFLDLK QGHYLPLNEA VYTRICSGAF Highest antigen sequence indentity to the following orthologs: Rat - 83%, Mouse - 80%.
SLC27A1 is a protein coding gene. Mediates the ATP-dependent import of long-chain fatty acids (LCFA) into the cell by mediating their translocation at the plasma membrane. Has also an acyl-CoA ligase activity for long-chain and very-long-chain fatty acids. May act directly as a bona fide transporter, or alternatively, in a cytoplasmic or membrane-associated multimeric protein complex to trap and draw fatty acids towards accumulation. Plays a pivotal role in regulating available LCFA substrates from exogenous sources in tissues undergoing high levels of beta-oxidation or triglyceride synthesis. May be involved in regulation of cholesterol metabolism.
Specifica
| SLC27A1 | |
| Polyclonal | |
| Unconjugated | |
| SLC27A1 | |
| ACSVL5; ACSVL5FATP-1; EC 6.2.1; Fatp; FATP1; FATP-1; Fatty acid transport protein; fatty acid transport protein 1; FLJ00336; long-chain fatty acid transport protein 1; MGC71751; SLC27A1; solute carrier family 27 (fatty acid transporter), member 1; solute carrier family 27 member 1; solute carrier family 27, member 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 376497 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6PCB7 | |
| SLC27A1 | |
| Recombinant protein corresponding to Human SLC27A1. Recombinant protein control fragment (Product #RP-108661). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto