missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMAD1 Mouse anti-Human, Mouse, Rat, Clone: 2D10B8, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66559-1-IG-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The protein encoded by this gene belongs to the SMAD, a family of proteins similar to the gene products of the Drosophila gene ′mothers against decapentaplegic′ (Mad) and the C. elegans gene Sma. SMAD proteins are signal transducers and transcriptional modulators that mediate multiple signaling pathways. This protein mediates the signals of the bone morphogenetic proteins (BMPs), which are involved in a range of biological activities including cell growth, apoptosis, morphogenesis, development and immune responses. In response to BMP ligands, this protein can be phosphorylated and activated by the BMP receptor kinase. The phosphorylated form of this protein forms a complex with SMAD4, which is important for its function in the transcription regulation. This protein is a target for SMAD-specific E3 ubiquitin ligases, such as SMURF1 and SMURF2, and undergoes ubiquitination and proteasome-mediated degradation. Alternatively spliced transcript variants encoding the same protein have been observed.Specifica
| SMAD1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P70340, P97588, Q15797 | |
| Smad1 | |
| SMAD1 Fusion Protein Ag0701 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2D10B8 | |
| Unconjugated | |
| SMAD1 | |
| BSP 1, BSP1, hSMAD1, JV4 1, JV41, MAD homolog 1, Mad related protein 1, MADH1, MADR1, Mothers against DPP homolog 1, SMAD 1, SMAD family member 1, SMAD1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 17125, 25671, 4086 | |
| -20°C | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto