Learn More
Small Sample Viscosity Standard ASTM D2162; UKAS ISO/IEC17025 and ISO 17034 certified, nominal kinematic viscosity 1009 cSt (40°C), Paragon Scientific™
Click to view available options
Quantità:
60 mL
Dimensione della confezione:
60mL
Descrizione
- Fully traceable to international standards
- Ensures full compliance to ASTM & IP test method protocol
- Certified in strict accordance with ASTM D2162
- 60mL lower volume cost effective solution
- Tamper evident security packaging
- 2 Year shelf life
- Manufactured in the United Kingdom
Specifica
Specifica
| Quantità | 60 mL |
Nominal values quoted. Certified Values will vary from batch to batch.