Learn More
Greiner Bio-One Small Volume™ 384 Deep Well Polypropylene Microplates
Click to view available options
Colore:
Black
Sterilità:
Non-sterile
Dimensione della confezione:
pezzi 100
Descrizione
- Non-pyrogenic
- Conical well design with flat bottom
- Ensures accurate pipetting
- Reduces dead volume of well
- Square well geometry, compatible with CapMat plate sealers
- Accommodates heat-sealing systems
Service Plan
No
Warranty
No
Specifica
Specifica
| Formato | Standard |
| Matrice | 16 x 24 |
| Colore | Black |
| Materiale | Polypropylene |
| No. of Wells | 384 |
| Sterilità | Non-sterile |
| Total Volume (Metric) Well | 1 to 90 μL |
| Design pozzetto | Deep well |