missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMO Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 20787-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Smoothened drosophila homolog (SMOH) is a serpentine protein belonging to the G-protein coupled receptor Fz/Smo family with 787 aa, seven transmembrane G protein-coupled receptors with 4 glycosylation sites and a putative extracellular amino terminus. Smoothened activated by b-Arrestin 2 and GRK2 acts as a receptor for Shh, the Sonic hedgehog gene product, and is a positive regulator of hedgehog signaling pathway along with the smoothened inhibitor patched protein (PTCH) and regulates vertebrate developmental processes, cellular proliferation and differentiation by activating transcription factor Gli. Smoothened mutants are known to cause sporadic basal cell carcinoma of the skin. It is known to express widely in heart and brain.Specifica
| SMO | |
| Polyclonal | |
| Unconjugated | |
| SMO | |
| Gx, Protein Gx, SMO, SMOH, Smoothened homolog | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6608 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.44 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q99835 | |
| SMO | |
| SMO Fusion Protein Ag14322 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto