Learn More
Motic™ SMZ161 Microscope
Click to view available options
Head Style:
Binocular
Trinocular
Tipo di illuminazione:
None
Quartz Halogen with Intensity Control 12V/10w
Quartz Halogen with Intensity Control 12V/20w
Dimensione della confezione:
Pezzo
Descrizione
- Greenough optical system.
- Sharp, three-dimensioned images and wide field of view.
- Interpupillary distance: 50-75mm.
- Ergonomically designed for long hours of viewing without discomfort.
- Suitable for industrial applications, assembly and inspection of small parts or integrated circuits on PCB, LCD production line.
- Coarse focusing system with tension adjustment.
Specifica
Specifica
| Include | Black and white stage plates and dust cover |
| Stile della base | Pole-type stand with transmitted illumination, 237x170mm |
| Potenza di ingrandimento | 0.75X to 4.5X |
| Tipi di obiettivo | Zoom system, ratio 1:6 |
| Intervallo di zoom | 1:6 |
| Product Type | Stereo zoom microscope |
| Modello | SMZ161-BL |
| Head Style | Binocular |
| Tipo di illuminazione | Quartz Halogen with Intensity Control 12V/20w |
| Eyepiece | WF10X/20 mm |