Learn More
SNN Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 13914-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The SNN antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SNN. This antibody recognizes human, mouse, rat antigen. The SNN antibody has been validated for the following applications: IHC, IF/ICC, ELISA analysis.Specifica
| SNN | |
| Polyclonal | |
| Unconjugated | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Polyclonal Antibody | |
| IgG |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA | |
| 450 μg/mL | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 8303 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |