missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 10304-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This endosomal protein regulates the cell-surface expression of epidermal growth factor receptor. This protein also has a role in sorting protease-activated receptor-1 from early endosomes to lysosomes. This protein may form oligomeric complexes with family members. This gene results in three transcript variants encoding distinct isoforms.Specifica
| SNX1 | |
| Polyclonal | |
| Unconjugated | |
| SNX1 | |
| HsT17379, SNX1, SNX1A, sorting nexin 1, Vps5 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 56440, 6642, 84471 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q13596, Q99N27, Q9WV80 | |
| Snx1 | |
| SNX1 Fusion Protein Ag0499 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto