missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Dodecyl Sulfate, MP Biomedicals
Sfoglia Tutto MP Biomedicals Prodotti
Click to view available options
Quantità:
25 g
500 g
1 kg
Dimensione della confezione:
1kg
25g
500g
Description
- Used to solubilize and denature proteins for denaturing-PAGE
- Most proteins bind SDS in a ratio of 1.4 g SDS per gram of protein
- The charges intrinsic to the protein become insignificant compared to the overall negative charge provided by the bound SDS
- The charge to mass ratio is essentially the same for each protein and will migrate in the gel based only on their size
Specifications
Specifications
| Materiale o nome chimico | SODIUM DODECYL SULFATE |
| Colore | White |
| CAS | 151-21-3 |
| Forma fisica | Powder |
| Punto di fusione | 180°C to 210°C |
| Quantità | 1 kg |
| Formula molecolare | C12H25NaO4S |
| Formula lineare | C12H25O4SNa |
| Numero MDL | MFCD00036175 |
| Sinonimo | SDS, Lauryl sulfate sodium salt, Dodecyl sulfate sodium salt, Dodecyl sodium sulfate, Sodium lauryl sulfate, Sulfuric acid monododecyl ester sodium salt |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction