missing translation for 'onlineSavingsMsg'
Learn More

Sodium ethoxide, 21% w/w in ethanol

Codice prodotto. p-7046084
Click to view available options
Quantità:
100 mL
500 mL
2500 mL
Dimensione della confezione:
100mL
2.50L
500mL
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Codice prodotto. 11337876

Marca: Thermo Scientific Alfa Aesar L13083.AE

Richiesta di formato massivo o personalizzato

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 141-52-6 | C2H5NaO | 68.051 g/mol

Sodium ethoxide, 21% w/w in ethanol is used as a strong base in organic synthesis. It finds application in various chemical reactions such as condensation, esterification, alkoxylation and etherifcation. It is actively involved in Claisen condensation, Stobbe reaction and Wolf-kishner reduction. It is an important starting material for the synthesis of ethyl ester and diethyl ester of malonic acid. In Williamson ether synthesis, it reacts with ethyl bromide to form diethyl ether.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium ethoxide, 21% w/w in ethanol is used as a strong base in organic synthesis. It finds application in various chemical reactions such as condensation, esterification, alkoxylation and etherifcation. It is actively involved in Claisen condensation, Stobbe reaction and Wolf-kishner reduction. It is an important starting material for the synthesis of ethyl ester and diethyl ester of malonic acid. In Williamson ether synthesis, it reacts with ethyl bromide to form diethyl ether.

Solubility
Miscible with alcohol.

Notes
Moisture sensitive. It hydrolyzes rapidly into sodium hydroxide in the presence of moist air. Incompatible with water, strong bases, acids, oxidizing agents, alkali metals, strong oxidizing agents, ammonia, acid chlorides, acid anhydrides, reducing agents, peroxides, acids and chlorinated solvents.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 141-52-6
Formula molecolare C2H5NaO
Molecular Weight (g/mol) 68.051
Numero MDL MFCD00012417
InChI Key QDRKDTQENPPHOJ-UHFFFAOYSA-N
Sinonimo sodium ethoxide, sodium ethylate, sodium ethanolate, sodiumethoxide, ethoxysodium, ethanol, sodium salt, caustic alcohol, naoet, etona, ethanol, sodium salt 1:1
PubChem CID 2723922
ChEBI CHEBI:52096
IUPAC Name sodio; etanolato
SMILES CC[O-].[Na+]

Specifica

Materiale o nome chimico Sodium ethoxide
Densità 0.868
Punto d'infiammabilità 20°C (68°F)
Formula lineare NaOCH2CH3
Indice di rifrazione 1.3875
Quantità 100 mL
Numero UN UN3274
Beilstein 3593646
Sensibilità Air and moisture sensitive
Indice di Merck 14,8613
Informazioni di solubilità Miscible with alcohol.
Peso formulazione 68.05
Concentration or Composition (by Analyte or Components) 21% w/w in ethanol
Vedi altri risultati Mostra meno risultati

For Manufacturing and Laboratory Use. Not for beverage use

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato