Learn More
Sodium phosphate, 0.2M buffer soln., pH 7.6, Thermo Scientific™
Click to view available options
Quantità:
500 mL
1 L
Dimensione della confezione:
1L
500mL
Descrizione
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifica
Specifica
| Materiale o nome chimico | Sodium phosphate buffer soln. |
| Concentrazione | 0.2 M |
| pH | 7.6 |
| Forma fisica | Liquid |
| Quantità | 1 L |
| Numero MDL | MFCD00146206 |