Learn More
Thermo Scientific Chemicals Sodium phosphate, 0.5M buffer soln., pH 7.0
Codice prodotto. 15485959
Click to view available options
Quantità:
250 mL
500 mL
Dimensione della confezione:
250mL
500mL
Codice prodotto. 15485959
Marca: Thermo Scientific Chemicals J63791.AP
Richiesta di formato massivo o personalizzato
Sodium phosphate is widely used in molecular biology, biochemistry, and chromatography. Useful in conjunction with Sodium Phosphate, Monobasic in the preparation of biological buffers. Used in many applications including the purification of antibodies.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium phosphate is widely used in molecular biology, biochemistry, and chromatography. Useful in conjunction with Sodium Phosphate, Monobasic in the preparation of biological buffers. Used in many applications including the purification of antibodies.
Solubility
Fully miscible.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico Sodium phosphate buffer soln.
Concentrazione 0.5 M
pH 7.0
Forma fisica Liquid
Quantità 500 mL
Numero MDL MFCD00146206
Informazioni di solubilità Fully miscible.