Learn More
Thermo Scientific Chemicals Sodium phosphate, 0.5M buffer soln., pH 8.5
Click to view available options
Quantità:
250 mL
500 mL
Dimensione della confezione:
250mL
500mL
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Materiale o nome chimico | Sodium phosphate buffer soln. |
| Concentrazione | 0.5 M |
| pH | 8.5 |
| Forma fisica | Liquid |
| Quantità | 250 mL |
| Numero MDL | MFCD00146206 |