Learn More
Memmert™ Software AtmoCONTROL for SN55plus
Sfoglia Tutto Memmert Prodotti
Click to view available options
Da utilizzare con (apparecchiatura):
SN55plus Sterilizer
Dimensione della confezione:
Pezzo
Descrizione
- Software conforming to FDA AtmoCONTROL
- Meets the requirements for the use of electronically stored data sets and electronic signatures as laid down in Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA)
- Base licence for the control of one unit
- Respective IQ/OQ documents available in German and English language (without surcharge)
Specifica
Specifica
| Certificazione/Conformità | Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA) |
| Da utilizzare con (apparecchiatura) | SN55plus Sterilizer |
| Product Type | Software |