Learn More
Brady™ Brass Padlocks
Click to view available options
Colore:
Black
Blue
Gray
Green
Red
Yellow
Dimensione della confezione:
1 BX
pezzi 12
Descrizione
- 6-pin cylinder offers more unique key cuts for better tamper resistance
- Locks are protected by coloured vinyl sheaths making it easy to color-code your lockout program
- Padlocks are available keyed different
- Lock labels sold separately
- Two keys supplied with each padlock
Specifica
Specifica
| Product Type | Brass Padlocks |
| Alternate Part Number | 51354 |
| Colore | Red |
| Materiale | Brass |
| Descrizione | Brass Padlocks |
| Diametro (sistema metrico) maniglione | 6.5 mm |
| Dimensioni (L x P) | 43.00 x 60.00 mm |
| Altezza (sistema metrico) | 21 mm |
| Altezza maniglione (sistema metrico) | 21.00 mm |
| Material (Shackle) | Brass |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion