Learn More
AMPri™ SolidSafety ChemN Special Nitrile Gloves
Click to view available options
Size:
Large
X-Large
Dimensione della confezione:
pezzi 100
Descrizione
- Category III
- Medical device Class I
- Food grade
- Chemical and physical resistance
- Proven resistance to microorganisms
- Silicone-free
- Soft, comfortable fit, wearing comfort
- Vera good grip, even if used in damp or wet environments
- Non-sterile
- Powder-free
- Color: green
Specifica
Specifica
| Certifications/Compliance (Europe) | EN 420, EN 455, EN 374/Type B (JKPT), Food grade according to Regulation (EU) 1935/2004, PPE Category III, Medical device Class I according to Regulation (EU) 2016/745 |
| AQL | 0.65 |
| Colore | Green |
| Materiale | Nitrile |
| Powdered | Powder Free |
| Size | Large |
| Lunghezza (sistema metrico) | 240 mm |