Learn More
Tocris Bioscience™ Spadin
Click to view available options
Quantità:
1 mg
Dimensione della confezione:
1mg
Descrizione
Spadin is a potent TREK-1 channel blocker (IC50 = 71 nM). Enhances dorsal raphe nucleus 5-HT neurotransmission in mice. Induces hippocampal CREB activation and neurogenesis in adult mice. Exhibits antidepressant effects in mouse models of depression. Brain penetrant.
Specifica
Specifica
| Purezza | 0.95 |
| Descrizione | Spadin |
| Peso molecolare | 2012.34 |
| Product Type | Spadin |
| Quantità | 1 mg |
| Sequenza | Tyr-Ala-Pro-Leu-Pro-Arg-Trp-Ser-Gly-Pro-Ile-Gly-Val-Ser-Trp-Gly-Leu-Arg |
For Research Use Only