Learn More
Spectrum™ Micro Float-A-Lyzer™ Ready-to-Use Small Volume Dialysis Device
Codice prodotto. 12085625
Click to view available options
MWCO:
0.1 to 0.5 kDa
0.5 to 1 kDa
100 kDa
20 kDa
3.5 to 5 kDa
50 kDa
8 to 10 kDa
Colore chiusura:
Black
Blue
Green
Orange
Red
White
Yellow
Altezza (sistema metrico):
44 mm
63 mm
Dimensione della confezione:
pezzi 12
Codice prodotto. 12085625
Marca: Spectrum™ F235057
Providing ultimate convenience, this preassembled device incorporates Spectrum's ultrapure Biotech Grade Cellulose Ester (CE) membrane tubing into an easy-to-use design making laboroatory microdialysis easier than ever
Ready-to-Use and disposable, the Float-A-Lyzer G2 incorporates Biotech Grade Cellulose Ester (CE) membrane tubing into a preasembled dialysis device that is easy-to-use & provides ultimate convenience. Available in volume sizes: 1, 5, and 10 mL. The resealable screw cap is color-coded for 9 available MWCO's and provides open access for easy loading and total sample recovery with a disposable pipet (included for 5 and 10mL sizes). The flotation ring (included) provides buoyancy to maximize dialysis efficiency.
  • Designed to stand upright on bench-tops during experimental preparation as well as float and self-orient during dialysis
  • Leak-proof Luer-Lok™ cap provides easy access with the included syringe for easy loading and sample retrieval
  • Units can also interlock for the simultaneous dialysis of multiple samples
  • Ready-to-use and complete
  • Easy syringe loading (no needle)
  • Total sample recovery
  • Ultrapure Biotech CE (no cleaning)
  • Largest range of MWCOs
  • Maximizes SA & minimizes hold-up volume
Specifica
Product Type Micro Float-A-Lyzer Dialysis Device
MWCO 20 kDa
Membrana Cellulose Ester
Materiale Polycarbonate
Diametro (sistema imperiale) membrana 0.25 in.
Diametro (sistema metrico) membrana 6.4 mm
Altezza (sistema imperiale) 1.7 in.
Altezza (sistema metrico) 44 mm
Larghezza (sistema imperiale) 1.77 in.
Larghezza (sistema metrico) 45 mm
Capacità (sistema metrico) 100 to 200 μL
Colore chiusura Red
Materiale chiusura Polypropylene
Tipo di chiusura Luer lok
Contenuto kit 1mL syringe
Quantità Each
Da utilizzare con (applicazione) Designed to stand upright on bench-tops during experimental preparation as well as float and self-orient during dialysis