Learn More
Spectrum™ Tubo per dialisi pre-impregnato Spectra/Por™ 6 standard RC, 1-50 kD MWCO
Codice prodotto. 11405879
Click to view available options
Diametro (sistema metrico):
7.5 mm
11.5 mm
15 mm
17.8 mm
20.4 mm
21.6 mm
24 mm
29 mm
31.8 mm
34.4 mm
MWCO:
1 kDa
10 kDa
15 kDa
2 kDa
25 kDa
3.5 kDa
50 kDa
8 kDa
Dimensione della confezione:
Pezzo
Codice prodotto. 11405879
Marca: Spectrum™ 132594
Clear, flexible and sturdy; Standard Grade RC (regenerated cellulose) membrane conveniently pre-equilibrated in water, eliminating the need to soak to remove glycerin and hydrate the membrane
This ready-to-use membrane is packaged wet in a preservative solution of 0.05% sodium azide. Simply rinse away the preservative solution with DI water, load the sample and dialyze.
  • Pre-wetted & ready-to-use, no glycerin removal required
  • 8 MWCO's ranging from 1 kD - 50 kD
  • Good chemical compatibility with dilute strong acids and bases, conc. weak acids & bases, many alcohols and some organics
  • Standard RC can tolerate pH 2 - 12 and temp 4-121°C.
  • Ideal for sample prep, desalting, buffer exchange, pH change, protein purification, etc.
  • Economic 10 meters/pkg
Do not tie knots in tubing; use closures
Specifica
Product Type Spectra/Por 6 Dialysis Tubing
MWCO 3.5 kDa
Materiale Regenerated Cellulose
Diametro (sistema metrico) 34.4 mm
Diametro (sistema imperiale) 1.35 in.
Lunghezza (sistema imperiale) 33 ft.
Lunghezza (sistema metrico) 10 m
Formulazione Packaged wet in 0.05% sodium azide
Intervallo di temperatura (sistema imperiale) 39.2°F to 249.8°F
Temperature Range (Metric) 4°C to 121°C
Quantità Each
Da utilizzare con (applicazione) Ideal for molecular separation, protein binding studies, electrolyte removal, desalting