Learn More
Spectrum™ Trial Size Kits for Biotech-Grade CE Dialysis Membrane Tubing (1 meter), MWCO: 300 kd
Codice prodotto. p-4394885
Click to view available options
Diametro (sistema metrico):
10 mm
20 mm
Dimensione della confezione:
1m
Pezzo
Codice prodotto. 15340682
Marca: Spectrum™ 131456T
Includes
  • 1m Biotech CE Tubing in 0.05% sodium azide
  • Two universal tubing closures
  • Five opening picks
Ideal for small, critical dialysis applications that require the more concise selectivity and retention provided by Biotech Cellulose Ester (CE) Membrane than Standard Grade RC
Kits are great for comparing membrane MWCO performance to optimize separation or purification. Provides a 1m length of Biotech-Grade CE membrane tubing (16mm flat width), a pair of universal tubing closures and 5 picks for opening membrane tubing. Tubing is wet packaged in 0.05% sodium azide preservative that easily rinses out with water. Nine different MWCOs avilalable: 0.1-0.5 kD, 0.5-1.0 kD, 3.5-5 kD, 8-10 kD, 20 kD, 50 kD, 100 kD, 300 kD and 1000 kD.
  • Rigidly controlled porosity for better selectivity and separations than Standard Grade RC
  • 9 MWCOs available ranging from 100 to 1,000,000 Daltons
  • Ultrapure, no heavy metal or sulfiide impurities
  • Low protein binding
  • No glycerin removal, cleaning or pre-treatment required
Specifica
MWCO 300 kDa
Diametro (sistema metrico) 20 mm
Lunghezza (sistema metrico) 3.1 mL/cm vol./length
Quantità 1 m Tubing/Kit