Learn More
Cole-Parmer™ Vial Extractor/End Plug Remover for Mid-Size and Large Grinding Vials
Codice prodotto. 15370492 Sfoglia Tutto SPEX Certiprep Prodotti
Click to view available options
:
Vial Extractor/End Plug Remover; For use with: Grinding Vial Set (Large, Mid Size), Cr-Free Grinding Vial (Large, Mid Size) of Spex SamplePrep Large-Capacity Freezer/Mill
Codice prodotto. 15370492
Marca: Cole-Parmer™ 6804
Questo articolo richiede un aggiornamento del prezzo prima di essere ordinato.
Richiedi qui il prezzo per questo articolo.
This Vial Extractor/End Plug Remover lifts large and mid-size grinding vials in and out of large Freezer/Mill.
  • Makes it easier to open cold grinding vials immediately after they have been removed from 6870 Large Freezer/Mill.
  • Lever helps to remove end plugs from center cylinders of large and mid-size grinding vials.
  • Compatible with following large grinding vials: 6801 Large Grinding Vial Set and 6871 Large low-chrome Grinding Vial Set
  • Compatible with following mid-size vials when used in conjunction with the 6884 Mid-Size Grinding Vial Adapter: 6881 Mid-Size Grinding Vial Set, 6883 Mid-Size low-chrome Grinding Vial Set and 6885 Mid-Size Poly-Vial Set
  • Included in 6870L Large Grinding Vial Accessory Package.
Specifica
Product Type Replacement Extraction Tool
Da utilizzare con (applicazione) Remove End Plugs From The Center Cylinders of Large and Mid Size Grinding Vials
Da utilizzare con (apparecchiatura) Grinding Vial Set (Large, Mid Size), Cr-Free Grinding Vial (Large, Mid Size) of Spex SamplePrep Large-Capacity Freezer/Mill