Learn More
Tygon SPT-3350 Platinum-Cured Silicone Tubing
Sfoglia Tutto Cole-Parmer Prodotti
Click to view available options
Diametro (sistema imperiale) interno:
1 in.
1-1/2 in.
1-1/4 in.
1/16 in.
1/2 in.
1/32 in.
1/4 in.
1/8 in.
3/16 in.
3/32 in.
3/4 in.
3/8 in.
5/16 in.
5/32 in.
5/8 in.
7/16 in.
Diametro (sistema imperiale) esterno:
1 in.
1-1/2 in.
1-1/4 in.
1/2 in.
1/4 in.
1/8 in.
11/16 in.
13/16 in.
2 in.
3/16 in.
3/32 in.
3/4 in.
3/8 in.
5/16 in.
5/32 in.
5/8 in.
7/16 in.
7/32 in.
7/8 in.
9/16 in.
Dimensione della confezione:
15m
Descrizione
Ideal for food and beverage transfer. Minimal e in., tractable maintains fluid integrity. Up to three times smoother than other silicone tubing. FDA 21 CFR 175.300 compliant. Free technical application support available to assist with tubing selection
Specifica
Specifica
| Certificazione/Conformità | USP;NSF-51;FDA |
| Diametro (sistema imperiale) interno | 1/2 in. |
| Materiale | Tygon 3350 |
| Diametro (sistema imperiale) esterno | 5/8 in. |
| Overall Length (English) | 50 ft. |
| Metodo di sterilizzazione | Yes |
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.