Learn More
SR9243, MedChemExpress
Product Code. 16497228
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16497228 10 mg 10mg
16483247 50 mg 50mg
16479757 5 mg 5mg
16457206 100 mg 100mg
16451236 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16497228 Supplier Medchem Express Supplier No. HY16972
 Restricted Item
Spedizione stimata: 30-11-2026
MedChemExpress SR9243 is a liver-X-receptor (LXR) inverse agonist that induces LXR-corepressor interaction.
SR9243 is a liver-X-receptor (LXR) inverse agonist that induces LXR-corepressor interaction.
Specifications
Chemical Name or Material SR9243
CAS 1613028-81-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C31H32BrNO4S2
Quantity 10 mg
Solubility Information DMSO : 20 mg/mL (31.92 mM; Need ultrasonic)
SMILES BrC1=CC(CCN(S(C2=C(C)C=C(C)C=C2C)(=O)=O)CC(C=C3)=CC=C3C4=CC(S(=O)(C)=O)=CC=C4)=CC=C1
Molecular Weight (g/mol) 626.62
Formula Weight 626.62
Percent Purity 95.03%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only