Learn More
SSBP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12212-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This protein binds preferentially and cooperatively to ss-DNA. Probably involved in mitochondrial DNA replication. SSBP is a housekeeping gene involved in mitochondrial biogenesis (Tiranti et al. 1995).Specifica
| SSBP1 | |
| Polyclonal | |
| Unconjugated | |
| SSBP1 | |
| Mt SSB, MtSSB, PWP1 interacting protein 17, SSBP, SSBP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 381760, 54304, 6742 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Western Blot, Immunocytochemistry | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P28042, Q04837, Q9CYR0 | |
| Ssbp1 | |
| SSBP1 Fusion Protein Ag2860 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |