missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD15 Monoclonal Antibody (28)
Mouse Monoclonal Antibody
Marca: Invitrogen™ MA134583
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
MA1-34583 detects CD15 from human samples.
CD15 (Lewis X, Le(x); stage specific embryonic antigen-1, SSEA-1) is a trisacharide determinant (3-fucosyl-N-acetyllactosamine) expressed on several glycolipids, glycoproteins and proteoglycans of various cell types, e.g. granulocytes, mast cells, monocytes, macrophages, cells of gastric mucosa, nervous system or various tumour cells. There are several variants of Lewis x, such as sialyl-Lewis x or sulphated Lewis x. Cells with high surface expression of Le(x) antigen exhibit strong self-aggregation, based on calcium-dependent Le(x)-Le(x) interaction. This process is involved for example in embryo compaction or in autoaggregation of teratocarcinoma cells. Sialyl-Le(x) and its isomer sialyl-Le(a) are ligands of selectins. CD15 expression has been extensively used to confirm diagnosis of Hodgkin's disease.
Specifica
| CD15 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P22083 | |
| FUT4 | |
| Cell preparation: Monocytes, separated from other peripheral blood leucocytes (PBL) on fibronectin plates. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgM |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Immunoprecipitation | |
| 28 | |
| Unconjugated | |
| FUT4 | |
| FUTIV; Lewis X; SSEA-1 | |
| Mouse | |
| IgG fraction | |
| RUO | |
| 2526 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto