Learn More
Labconco Stackable Scrubber Stand
Sfoglia Tutto Labconco Corporation Prodotti
Click to view available options
:
Stand: Glassware Washers; Type: Freestanding
Descrizione
- Reduced footprint compared to two undercounter or freestanding units
- Increased washing capacity with two Labconco Glassware Washers
- Strong and stable to support up to 4000 lb.
- Durable materials such as 1.75 in. tubular, Glacier White powder-coated steel
- Hardware included to securely mount top washer to stand
Specifica
Specifica
| Materiale | Powder-coated Steel |
| Product Type | Base Stand |
| Colore | White |